Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTGTGGTGGAGCTGGGGACACG[A/C]TGGGTAGGATGGTCAGGGAAAGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138250 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH18A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ALDH18A1 - aldehyde dehydrogenase 18 family member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017423.1 | Intron | NP_001017423.1 | ||||
NM_001323412.1 | Intron | NP_001310341.1 | ||||
NM_001323413.1 | Intron | NP_001310342.1 | ||||
NM_001323414.1 | Intron | NP_001310343.1 | ||||
NM_001323415.1 | Intron | NP_001310344.1 | ||||
NM_001323416.1 | Intron | NP_001310345.1 | ||||
NM_001323417.1 | Intron | NP_001310346.1 | ||||
NM_001323418.1 | Intron | NP_001310347.1 | ||||
NM_001323419.1 | Intron | NP_001310348.1 | ||||
NM_002860.3 | Intron | NP_002851.2 |
Set Membership: |
HapMap |