Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAATGTTTGTAATGAGAGATCAA[A/G]TGATTTCAGATAATTAAAAAAAATA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606160 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PANK1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PANK1 - pantothenate kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138316.3 | 4008 | UTR 3 | NP_612189.2 | |||
NM_148977.2 | 4008 | UTR 3 | NP_683878.1 | |||
NM_148978.2 | 4008 | UTR 3 | NP_683879.1 | |||
XM_017016333.1 | 4008 | Intron | XP_016871822.1 | |||
XM_017016334.1 | 4008 | Intron | XP_016871823.1 | |||
XM_017016335.1 | 4008 | Intron | XP_016871824.1 | |||
XM_017016336.1 | 4008 | Intron | XP_016871825.1 | |||
XM_017016337.1 | 4008 | Intron | XP_016871826.1 | |||
XM_017016338.1 | 4008 | Intron | XP_016871827.1 |
Set Membership: |
HapMap |