Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATATAAGTGTGAGTCAATCACTA[G/T]ACAGAATTTCATGTGTATGGATTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606115 | ||||||||||||||||||||
Literature Links: |
RPP30 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RPP30 - ribonuclease P/MRP subunit p30 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001104546.1 | Intron | NP_001098016.1 | ||||
NM_006413.4 | Intron | NP_006404.1 | ||||
XM_011539120.2 | Intron | XP_011537422.1 | ||||
XM_011539121.2 | Intron | XP_011537423.1 | ||||
XM_017015475.1 | Intron | XP_016870964.1 | ||||
XM_017015476.1 | Intron | XP_016870965.1 | ||||
XM_017015477.1 | Intron | XP_016870966.1 | ||||
XM_017015478.1 | Intron | XP_016870967.1 |