Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTACTGACCGCCCAGAGGTGGTGGA[A/G]TCTCTCTTCCCACAATGGCTGTTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612302 | ||||||||||||||||||||
Literature Links: |
ADGRA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADGRA1 - adhesion G protein-coupled receptor A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083909.2 | Intron | NP_001077378.1 | ||||
NM_001291085.1 | Intron | NP_001278014.1 | ||||
XM_005252695.1 | Intron | XP_005252752.1 | ||||
XM_011540273.1 | Intron | XP_011538575.1 | ||||
XM_017016779.1 | Intron | XP_016872268.1 |