Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTTTCAGGAGTTCTGAAAGCAA[A/C]CAACTGTTAAATAAACAGAAAAATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614649 MIM: 609520 | ||||||||||||||||||||
Literature Links: |
RNF170 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RNF170 - ring finger protein 170 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160223.1 | Intron | NP_001153695.1 | ||||
NM_001160224.1 | Intron | NP_001153696.1 | ||||
NM_001160225.1 | Intron | NP_001153697.1 | ||||
NM_030954.3 | Intron | NP_112216.3 | ||||
XM_006716404.2 | Intron | XP_006716467.1 | ||||
XM_006716405.2 | Intron | XP_006716468.1 | ||||
XM_011544666.2 | Intron | XP_011542968.1 | ||||
XM_017013881.1 | Intron | XP_016869370.1 | ||||
XM_017013882.1 | Intron | XP_016869371.1 |
THAP1 - THAP domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |