Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CACAGAATTCCCCCAAACAATTGTT[A/T]GGTATTTTTGTCGACTTTATTTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608736 | ||||||||||||||||||||
Literature Links: |
SLC39A14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC39A14 - solute carrier family 39 member 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128431.2 | Intron | NP_001121903.1 | ||||
NM_001135153.1 | Intron | NP_001128625.1 | ||||
NM_001135154.1 | Intron | NP_001128626.1 | ||||
NM_015359.4 | Intron | NP_056174.2 | ||||
XM_005273465.2 | Intron | XP_005273522.1 | ||||
XM_005273466.4 | Intron | XP_005273523.1 | ||||
XM_006716323.2 | Intron | XP_006716386.1 | ||||
XM_006716324.2 | Intron | XP_006716387.1 | ||||
XM_011544478.2 | Intron | XP_011542780.1 | ||||
XM_017013292.1 | Intron | XP_016868781.1 | ||||
XM_017013293.1 | Intron | XP_016868782.1 | ||||
XM_017013294.1 | Intron | XP_016868783.1 | ||||
XM_017013295.1 | Intron | XP_016868784.1 |