Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL18 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KLHL18 - kelch like family member 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025010.4 | Intron | NP_079286.2 | ||||
XM_005265001.3 | Intron | XP_005265058.2 | ||||
XM_005265002.4 | Intron | XP_005265059.2 | ||||
XM_005265003.3 | Intron | XP_005265060.2 | ||||
XM_006713079.1 | Intron | XP_006713142.1 | ||||
XM_017006033.1 | Intron | XP_016861522.1 | ||||
XM_017006034.1 | Intron | XP_016861523.1 | ||||
XM_017006035.1 | Intron | XP_016861524.1 |
Set Membership: |
HapMap |