Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGAGCACCACTCCAGCTGCCCC[C/T]TGGGCCGCACTCTCCCCCAGGCTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607071 MIM: 604038 MIM: 602725 MIM: 607073 | ||||||||||||||||||||
Literature Links: |
HYAL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HYAL1 - hyaluronoglucosaminidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033159.3 | 1777 | Silent Mutation | NP_149349.2 | |||
NM_153281.1 | 1777 | Silent Mutation | NP_695013.1 | |||
NM_153282.2 | 1777 | Intron | NP_695014.1 | |||
NM_153283.2 | 1777 | Silent Mutation | NP_695015.1 | |||
NM_153285.2 | 1777 | Silent Mutation | NP_695017.1 | |||
XM_011533667.2 | 1777 | Missense Mutation | XP_011531969.1 | |||
XM_011533668.2 | 1777 | Missense Mutation | XP_011531970.1 | |||
XM_011533669.2 | 1777 | Silent Mutation | XP_011531971.1 |
HYAL3 - hyaluronoglucosaminidase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IFRD2 - interferon related developmental regulator 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NAT6 - N-acetyltransferase 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |