Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAGATAAACGAGTTACATAATTG[G/T]CATAGAAAAAGGCAAATGCTACGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606112 | ||||||||||||||||||||
Literature Links: |
RPP14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RPP14 - ribonuclease P/MRP subunit p14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098783.2 | Intron | NP_001092253.1 | ||||
NM_007042.4 | Intron | NP_008973.1 | ||||
XM_011533313.2 | Intron | XP_011531615.1 | ||||
XM_017005641.1 | Intron | XP_016861130.1 |