Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACACTCCGAGTCTGCACTGGACC[A/G]TTCTCAGAGCGCTGTCCTGACTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601814 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FXYD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FXYD2 - FXYD domain containing ion transport regulator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001680.4 | Intron | NP_001671.2 | ||||
NM_021603.3 | Intron | NP_067614.1 |
FXYD6-FXYD2 - FXYD6-FXYD2 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204268.1 | Intron | NP_001191197.1 | ||||
NM_001243598.2 | Intron | NP_001230527.1 |
Set Membership: |
HapMap |