Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAGATCTGAGGTGCATAGAGTGT[A/C]GGGATTAAAAGTAGTTCTGGCTGAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 104240 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ST3GAL4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ST3GAL4 - ST3 beta-galactoside alpha-2,3-sialyltransferase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001254757.1 | Intron | NP_001241686.1 | ||||
NM_001254758.1 | Intron | NP_001241687.1 | ||||
NM_001254759.1 | Intron | NP_001241688.1 | ||||
NM_006278.2 | Intron | NP_006269.1 | ||||
XM_011542959.1 | Intron | XP_011541261.1 | ||||
XM_011542960.2 | Intron | XP_011541262.1 | ||||
XM_017018134.1 | Intron | XP_016873623.1 |
Set Membership: |
HapMap |