Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTTTGTGTTACAAACATTCCAA[C/T]TGCATTCTGATGAAATGTCAGACTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610110 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANO3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ANO3 - anoctamin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001313726.1 | Intron | NP_001300655.1 | ||||
NM_001313727.1 | Intron | NP_001300656.1 | ||||
NM_031418.3 | Intron | NP_113606.2 | ||||
XM_011520282.2 | Intron | XP_011518584.1 | ||||
XM_017018118.1 | Intron | XP_016873607.1 | ||||
XM_017018119.1 | Intron | XP_016873608.1 |