Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATAATTTTTCTCCTGAAGGCTTCA[A/G]ACATGAGCACGCTTCCAACTCATGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610110 | ||||||||||||||||||||
Literature Links: |
ANO3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANO3 - anoctamin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001313726.1 | 104 | Missense Mutation | AAA,AGA | K,R 17 | NP_001300655.1 | |
NM_001313727.1 | 104 | Intron | NP_001300656.1 | |||
NM_031418.3 | 104 | Intron | NP_113606.2 | |||
XM_011520282.2 | 104 | Intron | XP_011518584.1 | |||
XM_017018118.1 | 104 | Intron | XP_016873607.1 | |||
XM_017018119.1 | 104 | Intron | XP_016873608.1 |