Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCATGGCCACCTCCCCGCAGAAGT[A/C]GCCTTCTGTCCCCAAGTCTCCCACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608120 | ||||||||||||||||||||
Literature Links: |
PARVA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PARVA - parvin alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018222.4 | 192 | Nonsense Mutation | TAG,TCG | *,S 48 | NP_060692.2 | |
XM_005253015.3 | 192 | Intron | XP_005253072.1 |