Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGGCTCTCAACACCAACCCTGAC[A/G]GTCCTACAGGCCACTTGCCATGGGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DENND5A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DENND5A - DENN domain containing 5A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243254.1 | Intron | NP_001230183.1 | ||||
NM_015213.3 | Intron | NP_056028.2 | ||||
XM_005252832.1 | Intron | XP_005252889.1 | ||||
XM_011519953.2 | Intron | XP_011518255.1 | ||||
XM_017017419.1 | Intron | XP_016872908.1 |