Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATTCTGTCATTCAGCCAATATTT[A/C]TTGAACCAGAGATTGTGCTAGGCAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118445 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCKBR PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CCKBR - cholecystokinin B receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318029.1 | Intron | NP_001304958.1 | ||||
NM_176875.3 | Intron | NP_795344.1 | ||||
XM_005253210.1 | Intron | XP_005253267.1 | ||||
XM_017018516.1 | Intron | XP_016874005.1 |
Set Membership: |
HapMap |