Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCCTAGCCAGGGCCCAACCTGG[G/T]TCTATTAATAACTAAGCAAACTAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF557 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF557 - zinc finger protein 557 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001044387.1 | 66 | UTR 5 | NP_001037852.1 | |||
NM_001044388.1 | 66 | UTR 5 | NP_001037853.1 | |||
NM_024341.2 | 66 | UTR 5 | NP_077317.2 |