Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATGCTGAAGGCGTCTTGTCTGCC[G/T]CTCGGCTTCATCGTCTTCCTGCCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609156 MIM: 603751 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCLN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NCLN - nicalin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321463.1 | 211 | Silent Mutation | CCG,CCT | P,P 19 | NP_001308392.1 | |
NM_020170.3 | 211 | Silent Mutation | CCG,CCT | P,P 19 | NP_064555.2 |
S1PR4 - sphingosine-1-phosphate receptor 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |