Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCAGAAAAAAAACGATGTGTAAA[G/T]TATTGAAGTGATTGGAGAAGAATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
46 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609100 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO28 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FBXO28 - F-box protein 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136115.2 | Intron | NP_001129587.1 | ||||
NM_015176.3 | Intron | NP_055991.1 |