Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611367 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNPEP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DNPEP - aspartyl aminopeptidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319116.1 | Intron | NP_001306045.1 | ||||
NM_001319117.1 | Intron | NP_001306046.1 | ||||
NM_001319118.1 | Intron | NP_001306047.1 | ||||
NM_001319119.1 | Intron | NP_001306048.1 | ||||
NM_001319120.1 | Intron | NP_001306049.1 | ||||
NM_001319121.1 | Intron | NP_001306050.1 | ||||
NM_001319122.1 | Intron | NP_001306051.1 | ||||
NM_012100.3 | Intron | NP_036232.2 |
Set Membership: |
HapMap |