Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGGGTTACCAAAGGGGGATTCC[A/G]GGCAGGCAACGAATTGAGCGCTAGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
25 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC27 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC27 - coiled-coil domain containing 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152492.2 | Intron | NP_689705.2 | ||||
XM_011540780.1 | Intron | XP_011539082.1 | ||||
XM_017000402.1 | Intron | XP_016855891.1 | ||||
XM_017000403.1 | Intron | XP_016855892.1 | ||||
XM_017000404.1 | Intron | XP_016855893.1 | ||||
XM_017000405.1 | Intron | XP_016855894.1 | ||||
XM_017000406.1 | Intron | XP_016855895.1 |
TP73-AS1 - TP73 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |