Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACGGGGAGCCTCTCCCAGCATCCA[A/G]CCCCCCTTGCTCTGCCTCAGTCTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611251 | ||||||||||||||||||||
Literature Links: |
DISP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DISP3 - dispatched RND transporter family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020780.1 | Intron | NP_065831.1 | ||||
XM_011541828.2 | Intron | XP_011540130.1 | ||||
XM_011541829.1 | Intron | XP_011540131.1 | ||||
XM_011541830.2 | Intron | XP_011540132.1 | ||||
XM_011541831.2 | Intron | XP_011540133.1 | ||||
XM_011541832.2 | Intron | XP_011540134.1 | ||||
XM_011541833.2 | Intron | XP_011540135.1 | ||||
XM_011541835.2 | Intron | XP_011540137.1 |