Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGATGCTGAGAGAATTGTTTTAC[A/G]TATTTGTTAATCACGTCTCAGAATG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611251 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DISP3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DISP3 - dispatched RND transporter family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020780.1 | Intron | NP_065831.1 | ||||
XM_011541828.2 | Intron | XP_011540130.1 | ||||
XM_011541829.1 | Intron | XP_011540131.1 | ||||
XM_011541830.2 | Intron | XP_011540132.1 | ||||
XM_011541831.2 | Intron | XP_011540133.1 | ||||
XM_011541832.2 | Intron | XP_011540134.1 | ||||
XM_011541833.2 | Intron | XP_011540135.1 | ||||
XM_011541835.2 | Intron | XP_011540137.1 |