Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAATAAATAAAATAAAACGTCACT[C/T]TCACACTAATGCTGTCTAAGAGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616804 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
VSTM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
VSTM1 - V-set and transmembrane domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288791.1 | Intron | NP_001275720.1 | ||||
NM_001288792.1 | Intron | NP_001275721.1 | ||||
NM_001288793.1 | Intron | NP_001275722.1 | ||||
NM_198481.3 | Intron | NP_940883.2 | ||||
XM_011526845.1 | Intron | XP_011525147.1 | ||||
XM_011526846.1 | Intron | XP_011525148.1 | ||||
XM_011526847.1 | Intron | XP_011525149.1 | ||||
XM_011526848.1 | Intron | XP_011525150.1 | ||||
XM_011526849.1 | Intron | XP_011525151.1 | ||||
XM_017026666.1 | Intron | XP_016882155.1 |
Set Membership: |
HapMap |