Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCCGAAGTGATACAGTCTGAGAT[A/G]TTGAAAACATAGAAATTGGCCGGGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604530 MIM: 609661 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCR1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NCR1 - natural cytotoxicity triggering receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145457.2 | Intron | NP_001138929.2 | ||||
NM_001145458.2 | Intron | NP_001138930.2 | ||||
NM_001242356.2 | Intron | NP_001229285.1 | ||||
NM_001242357.2 | Intron | NP_001229286.1 | ||||
NM_004829.6 | Intron | NP_004820.2 | ||||
XM_011527528.2 | Intron | XP_011525830.1 | ||||
XM_011527529.2 | Intron | XP_011525831.1 | ||||
XM_011527530.2 | Intron | XP_011525832.1 |
NLRP7 - NLR family pyrin domain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127255.1 | Intron | NP_001120727.1 | ||||
NM_139176.3 | Intron | NP_631915.2 | ||||
NM_206828.3 | Intron | NP_996611.2 | ||||
XM_006723075.3 | Intron | XP_006723138.1 | ||||
XM_006723076.3 | Intron | XP_006723139.1 | ||||
XM_011526596.2 | Intron | XP_011524898.1 | ||||
XM_011526599.2 | Intron | XP_011524901.1 | ||||
XM_011526601.2 | Intron | XP_011524903.1 |
Set Membership: |
HapMap |