Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCAAGGATCTAGGAATTATACTC[C/T]CTCTAAATTGATTTATCCATCTTTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
22 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYDE2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SYDE2 - synapse defective Rho GTPase homolog 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032184.1 | Intron | NP_115560.1 | ||||
XM_005271254.3 | Intron | XP_005271311.1 | ||||
XM_006710946.3 | Intron | XP_006711009.1 | ||||
XM_017002483.1 | Intron | XP_016857972.1 | ||||
XM_017002484.1 | Intron | XP_016857973.1 | ||||
XM_017002485.1 | Intron | XP_016857974.1 | ||||
XM_017002486.1 | Intron | XP_016857975.1 |
Set Membership: |
HapMap |