Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGATTAAGGATAGAAGCAGTATAA[C/T]GTTCCATGGTGCTAATTCATGCTAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYDE2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SYDE2 - synapse defective Rho GTPase homolog 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032184.1 | Intron | NP_115560.1 | ||||
XM_005271254.3 | Intron | XP_005271311.1 | ||||
XM_006710946.3 | Intron | XP_006711009.1 | ||||
XM_017002483.1 | Intron | XP_016857972.1 | ||||
XM_017002484.1 | Intron | XP_016857973.1 | ||||
XM_017002485.1 | Intron | XP_016857974.1 | ||||
XM_017002486.1 | Intron | XP_016857975.1 |
Set Membership: |
HapMap |