Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACGTCACTGGGCCTTGCATGCTG[A/G]CCAGTAAGTGACCGCTGTTGACTCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608121 MIM: 608122 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PARVB PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PARVB - parvin beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PARVG - parvin gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001137605.2 | Intron | NP_001131077.1 | ||||
NM_022141.6 | Intron | NP_071424.1 | ||||
XM_005261702.3 | Intron | XP_005261759.1 | ||||
XM_011530302.2 | Intron | XP_011528604.1 | ||||
XM_017028907.1 | Intron | XP_016884396.1 | ||||
XM_017028908.1 | Intron | XP_016884397.1 |
Set Membership: |
HapMap |