Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATCAGGGTCCTTAATTCTCCAGG[A/C]AGAGCCTTAATCACTTTGCACCCTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102574 MIM: 603539 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTN3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ACTN3 - actinin alpha 3 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001104.3 | Intron | NP_001095.2 | ||||
NM_001258371.2 | Intron | NP_001245300.2 |
CTSF - cathepsin F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003793.3 | Intron | NP_003784.2 | ||||
XM_011545328.2 | Intron | XP_011543630.1 |
Set Membership: |
HapMap |