Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCTGGGCAGCAACCATTATGCAG[C/T]AGTGGCCTCCAAGCAGAAGCTGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603506 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRP5 - LDL receptor related protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291902.1 | Intron | NP_001278831.1 | ||||
NM_002335.3 | Intron | NP_002326.2 | ||||
XM_005273994.2 | Intron | XP_005274051.1 | ||||
XM_011545029.1 | Intron | XP_011543331.1 | ||||
XM_011545030.1 | Intron | XP_011543332.1 | ||||
XM_011545031.1 | Intron | XP_011543333.1 | ||||
XM_017017735.1 | Intron | XP_016873224.1 | ||||
XM_017017736.1 | Intron | XP_016873225.1 |