Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGTGCTTGGCACATAGCCCATAT[C/G]CATATATTTATTGAGTAAATGAGAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100133315 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC100133315 - transient receptor potential cation channel, subfamily C, member 2-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF121 - ring finger protein 121 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300926.1 | Intron | NP_001287855.1 | ||||
NM_018320.4 | Intron | NP_060790.2 | ||||
XM_006718629.1 | Intron | XP_006718692.1 | ||||
XM_006718630.1 | Intron | XP_006718693.1 | ||||
XM_006718632.1 | Intron | XP_006718695.1 | ||||
XM_011545150.1 | Intron | XP_011543452.1 |
Set Membership: |
HapMap |