Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGACATTCTGGGCTCTGTTAAT[C/G]TAAATGACTTCATGCCTACCCCCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616316 | ||||||||||||||||||||
Literature Links: |
FAM168A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM168A - family with sequence similarity 168 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286050.1 | Intron | NP_001272979.1 | ||||
NM_001286051.1 | Intron | NP_001272980.1 | ||||
NM_015159.2 | Intron | NP_055974.1 | ||||
XM_017017415.1 | Intron | XP_016872904.1 | ||||
XM_017017416.1 | Intron | XP_016872905.1 |