Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCATCCATTCCTCTGAAATACTCC[C/T]ACACTTCTGGCTGTAGACCTGAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615944 MIM: 602044 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2CD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C2CD3 - C2 calcium dependent domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286577.1 | Intron | NP_001273506.1 | ||||
NM_015531.5 | Intron | NP_056346.3 | ||||
XM_011544904.1 | Intron | XP_011543206.1 | ||||
XM_011544905.1 | Intron | XP_011543207.1 | ||||
XM_017017510.1 | Intron | XP_016872999.1 | ||||
XM_017017511.1 | Intron | XP_016873000.1 | ||||
XM_017017512.1 | Intron | XP_016873001.1 | ||||
XM_017017513.1 | Intron | XP_016873002.1 | ||||
XM_017017514.1 | Intron | XP_016873003.1 | ||||
XM_017017515.1 | Intron | XP_016873004.1 | ||||
XM_017017516.1 | Intron | XP_016873005.1 |
UCP3 - uncoupling protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |