Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAAACTATCTCAACAGCTTTCTC[C/T]TCTCCAGAGTCAAACAATCTTTATT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611610 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928580 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC101928580 - uncharacterized LOC101928580 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PGM2L1 - phosphoglucomutase 2 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173582.4 | 5460 | UTR 3 | NP_775853.2 | |||
XM_011544953.2 | 5460 | UTR 3 | XP_011543255.1 |
Set Membership: |
HapMap |