Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTCCGTTTCCCATGTGGCAGCCA[C/G]ATGGGGCTCCGCAAGACAGCCGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602537 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPN5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CAPN5 - calpain 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004055.4 | Intron | NP_004046.2 | ||||
XM_011545225.1 | Intron | XP_011543527.1 | ||||
XM_017018223.1 | Intron | XP_016873712.1 |
Set Membership: |
HapMap |