Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTTGAAGAAATAAAGACTAAAAT[A/T]ATGTCCAATTTGATGAAAATTATAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
GDPD4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GDPD4 - glycerophosphodiester phosphodiesterase domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182833.1 | 5512 | Intron | NP_878253.1 | |||
XM_011544826.1 | 5512 | Intron | XP_011543128.1 | |||
XM_011544833.1 | 5512 | Intron | XP_011543135.1 | |||
XM_011544834.1 | 5512 | Intron | XP_011543136.1 | |||
XM_017017348.1 | 5512 | UTR 3 | XP_016872837.1 | |||
XM_017017349.1 | 5512 | UTR 3 | XP_016872838.1 | |||
XM_017017350.1 | 5512 | UTR 3 | XP_016872839.1 | |||
XM_017017351.1 | 5512 | UTR 3 | XP_016872840.1 |