Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGAAATCTTCCAAGGGGCCTCAC[C/T]GCTTAGAGGATGAAATCTAAAATCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GDPD4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GDPD4 - glycerophosphodiester phosphodiesterase domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182833.1 | 3289 | Intron | NP_878253.1 | |||
XM_011544826.1 | 3289 | Intron | XP_011543128.1 | |||
XM_011544833.1 | 3289 | Intron | XP_011543135.1 | |||
XM_011544834.1 | 3289 | Intron | XP_011543136.1 | |||
XM_017017348.1 | 3289 | UTR 3 | XP_016872837.1 | |||
XM_017017349.1 | 3289 | UTR 3 | XP_016872838.1 | |||
XM_017017350.1 | 3289 | UTR 3 | XP_016872839.1 | |||
XM_017017351.1 | 3289 | UTR 3 | XP_016872840.1 |
Set Membership: |
HapMap |