Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTAACCCAACGCAGTGAAGGCTC[A/C]CAGTCTAGGAATTAAAGTGTTTCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606153 MIM: 612901 | ||||||||||||||||||||
Literature Links: |
ATP5E PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATP5E - ATP synthase, H+ transporting, mitochondrial F1 complex, epsilon subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006886.3 | Intron | NP_008817.1 |
PRELID3B - PRELI domain containing 3B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256403.1 | Intron | NP_001243332.1 | ||||
NM_016045.2 | Intron | NP_057129.2 |
SLMO2-ATP5E - SLMO2-ATP5E readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TUBB1 - tubulin beta 1 class VI | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |