Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CDH26 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDH26 - cadherin 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021810.4 | Intron | NP_068582.2 | ||||
NM_177980.2 | Intron | NP_817089.1 | ||||
XM_011528969.2 | Intron | XP_011527271.1 | ||||
XM_011528970.2 | Intron | XP_011527272.1 | ||||
XM_017027994.1 | Intron | XP_016883483.1 | ||||
XM_017027995.1 | Intron | XP_016883484.1 | ||||
XM_017027996.1 | Intron | XP_016883485.1 | ||||
XM_017027997.1 | Intron | XP_016883486.1 | ||||
XM_017027998.1 | Intron | XP_016883487.1 | ||||
XM_017027999.1 | Intron | XP_016883488.1 | ||||
XM_017028000.1 | Intron | XP_016883489.1 | ||||
XM_017028001.1 | Intron | XP_016883490.1 | ||||
XM_017028002.1 | Intron | XP_016883491.1 |