Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATCCTGGGCATTGATCCAAGTGC[C/G]AGGGTTAACATTTTTTTTTGAGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607343 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SALL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SALL4 - spalt like transcription factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318031.1 | Intron | NP_001304960.1 | ||||
NM_020436.4 | Intron | NP_065169.1 | ||||
XM_005260467.3 | Intron | XP_005260524.1 | ||||
XM_011528921.2 | Intron | XP_011527223.1 | ||||
XM_011528922.2 | Intron | XP_011527224.1 |
Set Membership: |
HapMap |