Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGTTTTATTTTCCTTTCTGGAAG[C/G]TTTTAGGGCCTTTATTTCTGGTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604032 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF2AK3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
EIF2AK3 - eukaryotic translation initiation factor 2 alpha kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001313915.1 | Intron | NP_001300844.1 | ||||
NM_004836.6 | Intron | NP_004827.4 | ||||
XM_017005376.1 | Intron | XP_016860865.1 |
LOC101928371 - uncharacterized LOC101928371 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |