Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTATCACCATTCACGCATTCTAC[A/G]CTTCATTATACGCTTCATCATAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
33 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFCAB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EFCAB2 - EF-hand calcium binding domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143943.1 | Intron | NP_001137415.1 | ||||
NM_001290327.1 | Intron | NP_001277256.1 | ||||
NM_032328.3 | Intron | NP_115704.1 | ||||
XM_011544300.2 | Intron | XP_011542602.1 | ||||
XM_011544301.1 | Intron | XP_011542603.1 | ||||
XM_011544302.2 | Intron | XP_011542604.1 | ||||
XM_017002538.1 | Intron | XP_016858027.1 | ||||
XM_017002539.1 | Intron | XP_016858028.1 | ||||
XM_017002540.1 | Intron | XP_016858029.1 | ||||
XM_017002541.1 | Intron | XP_016858030.1 | ||||
XM_017002542.1 | Intron | XP_016858031.1 | ||||
XM_017002543.1 | Intron | XP_016858032.1 | ||||
XM_017002544.1 | Intron | XP_016858033.1 | ||||
XM_017002545.1 | Intron | XP_016858034.1 |
Set Membership: |
HapMap |