Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTACCCAGCATCGCTGTCCTTTC[C/T]GTTTTTCTGTGGAGCTCTGGGCAGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
29 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR2T8 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
OR2T8 - olfactory receptor family 2 subfamily T member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005522.1 | 2098 | Intron | NP_001005522.1 | |||
XM_011544178.2 | 2098 | Intron | XP_011542480.1 | |||
XM_017001172.1 | 2098 | UTR 5 | XP_016856661.1 |
Set Membership: |
HapMap |