Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATATATATACTATATATTTTTTT[G/T]GTCAAATTTTATTTTATTATTATTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606208 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928414 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC101928414 - uncharacterized LOC101928414 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC28A2 - solute carrier family 28 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004212.3 | Intron | NP_004203.2 | ||||
XM_011522198.2 | Intron | XP_011520500.1 | ||||
XM_011522199.1 | Intron | XP_011520501.1 | ||||
XM_011522200.2 | Intron | XP_011520502.1 | ||||
XM_011522201.2 | Intron | XP_011520503.1 | ||||
XM_011522202.2 | Intron | XP_011520504.1 |
Set Membership: |
HapMap |