Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTTGTATACAATTTATACAATTT[A/T]ACGTATCTAGTGGTGGCTTAGAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610953 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PIF1 - PIF1 5'-to-3' DNA helicase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286496.1 | Intron | NP_001273425.1 | ||||
NM_001286497.1 | Intron | NP_001273426.1 | ||||
NM_001286499.1 | Intron | NP_001273428.1 | ||||
NM_025049.3 | Intron | NP_079325.2 | ||||
XM_011522083.2 | Intron | XP_011520385.1 | ||||
XM_011522084.2 | Intron | XP_011520386.1 |
Set Membership: |
HapMap |