Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCCTCCTTGGGCCTCTCCAGGT[C/T]ATGGAACACACTGTCCTCATGTTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614476 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CT62 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CT62 - cancer/testis antigen 62 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
THSD4 - thrombospondin type 1 domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286429.1 | Intron | NP_001273358.1 | ||||
NM_024817.2 | Intron | NP_079093.2 | ||||
XM_006720692.3 | Intron | XP_006720755.1 | ||||
XM_011522043.2 | Intron | XP_011520345.1 | ||||
XM_011522044.2 | Intron | XP_011520346.1 | ||||
XM_017022582.1 | Intron | XP_016878071.1 | ||||
XM_017022583.1 | Intron | XP_016878072.1 | ||||
XM_017022584.1 | Intron | XP_016878073.1 | ||||
XM_017022585.1 | Intron | XP_016878074.1 | ||||
XM_017022586.1 | Intron | XP_016878075.1 |