Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600374 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BBS4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BBS4 - Bardet-Biedl syndrome 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252678.1 | Intron | NP_001239607.1 | ||||
NM_001320665.1 | Intron | NP_001307594.1 | ||||
NM_033028.4 | Intron | NP_149017.2 | ||||
XM_011521848.1 | Intron | XP_011520150.1 | ||||
XM_011521849.1 | Intron | XP_011520151.1 | ||||
XM_011521851.1 | Intron | XP_011520153.1 | ||||
XM_017022450.1 | Intron | XP_016877939.1 | ||||
XM_017022451.1 | Intron | XP_016877940.1 | ||||
XM_017022452.1 | Intron | XP_016877941.1 | ||||
XM_017022453.1 | Intron | XP_016877942.1 | ||||
XM_017022454.1 | Intron | XP_016877943.1 |
HIGD2B - HIG1 hypoxia inducible domain family member 2B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |