Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGGCCCTGAAGTCACTGTTTTA[A/G]CTATCACACCCCTATCTTCACCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611861 | ||||||||||||||||||||
Literature Links: |
ADPGK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADPGK - ADP dependent glucokinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031284.4 | Intron | NP_112574.3 | ||||
XM_006720712.3 | Intron | XP_006720775.1 | ||||
XM_006720713.3 | Intron | XP_006720776.1 | ||||
XM_011522102.2 | Intron | XP_011520404.1 | ||||
XM_011522103.2 | Intron | XP_011520405.1 | ||||
XM_011522104.2 | Intron | XP_011520406.1 | ||||
XM_017022664.1 | Intron | XP_016878153.1 |