Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTTTTTGTTTTTTGTACTACTTA[A/G]TTTCAAAAGGACAAAATCCTTGGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611622 | ||||||||||||||||||||
Literature Links: |
IQCJ PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IQCJ - IQ motif containing J | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042705.2 | Intron | NP_001036170.1 | ||||
NM_001042706.2 | Intron | NP_001036171.1 | ||||
NM_001197100.1 | Intron | NP_001184029.1 |
IQCJ-SCHIP1 - IQCJ-SCHIP1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197113.1 | Intron | NP_001184042.1 | ||||
NM_001197114.1 | Intron | NP_001184043.1 |