Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608263 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSPBAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSPBAP1 - HSPB1 associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320728.1 | Intron | NP_001307657.1 | ||||
NM_024610.5 | Intron | NP_078886.2 | ||||
XM_005247762.2 | Intron | XP_005247819.1 | ||||
XM_011513130.2 | Intron | XP_011511432.1 | ||||
XM_011513132.2 | Intron | XP_011511434.2 | ||||
XM_017007179.1 | Intron | XP_016862668.1 | ||||
XM_017007180.1 | Intron | XP_016862669.1 |